Learn More
Bioworld Orchimax Medium w/ Charcoal, 25 L
Catalog No. 50254763
Encompass_Preferred
Orchimax MediumContains Macronutrients, Micronutrients, Activated Charcoal, Sucrose, Tryptone, and Vitamins MES is also included to prevent oxidation Cytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-763 Supplier Bioworld Supplier No. 306301043
Orchimax MediumContains Macronutrients, Micronutrients, Activated Charcoal, Sucrose, Tryptone, and Vitamins MES is also included to prevent oxidation Cytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested