Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Orcinol gentiobioside | 164991-86-0 | 5 MG
SDP

Catalog No. 5000420837
Encompass_Preferred

Orcinol gentiobioside is a natural product isolated from the rhizomes of *C. breviscapa*. This product is intended for research use only and is suitable for various compound screening libraries.

  • Purity of 99.0%
  • Molecular weight of 448.42
  • Chemical formula is C19H28O12
  • Appears as a white to off-white solid
  • Classified as a phenol, monophenol
  • Used in natural product, glycoside, medicine food homology, phenols, traditional Chinese medicine active, and plant-sourced natural product libraries

Catalog No. 50-004-20837 Supplier Medchemexpress LLC Supplier No. HYN41235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Orcinol gentiobioside is a natural product isolated from the rhizomes of *C. breviscapa*. This product is intended for research use only and is suitable for various compound screening libraries.

  • Purity of 99.0%
  • Molecular weight of 448.42
  • Chemical formula is C19H28O12
  • Appears as a white to off-white solid
  • Classified as a phenol, monophenol
  • Used in natural product, glycoside, medicine food homology, phenols, traditional Chinese medicine active, and plant-sourced natural product libraries

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.