Learn More
Cayman Chemical OrcInol GlucosIde 10mg
SDP
Catalog No. 502943491
Encompass_Preferred
A phenolic glycoside with diverse biological activities; scavenges hydrogen peroxide- and iron-induced hydroxyl- and superoxide anion radicals in cell-free assays (IC50s = 1.39 and 2.49 mM, respectively); increases proliferation, Alp activity, and levels of collagen I and Runx2 in primary mouse osteoblasts at 100 nM; prevents dexamethasone-induced decreases in femur bone mass density and volume in mice at 20 mg/kg per day; reduces immobility time in the forced swim test and serum levels of corticosterone in a rat model of chronic unpredictable stress-induced depression at 1.5, 3, or 6 mg/kg; increases the number of entries into and time spent in the open arms of the elevated plus maze in mice at 5, 10, or 20 mg/kg
Catalog No. 50-294-3491 Supplier Cayman Chemical Supplier No. 4236510 MG
A phenolic glycoside with diverse biological activities; scavenges hydrogen peroxide- and iron-induced hydroxyl- and superoxide anion radicals in cell-free assays (IC50s = 1.39 and 2.49 mM, respectively); increases proliferation, Alp activity, and levels of collagen I and Runx2 in primary mouse osteoblasts at 100 nM; prevents dexamethasone-induced decreases in femur bone mass density and volume in mice at 20 mg/kg per day; reduces immobility time in the forced swim test and serum levels of corticosterone in a rat model of chronic unpredictable stress-induced depression at 1.5, 3, or 6 mg/kg; increases the number of entries into and time spent in the open arms of the elevated plus maze in mice at 5, 10, or 20 mg/kg