Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Orcinol glucoside | 21082-33-7 | MFCD06794970 | 99.6% | 286.28 g·mol⁻¹ | C13H18O7 | 10 MG
SDP

Catalog No. 5000296782
Encompass_Preferred

Orcinol glucoside is a natural phenolic glycoside isolated from Curculigo orchioides and supplied as a research-grade analytical standard for in vitro studies. It modulates mesenchymal stem cell differentiation via Wnt/β-catenin signaling and is intended for research use only.

  • High purity: 99.57%
  • Molecular weight 286.28 g·mol⁻¹
  • Chemical formula C13H18O7
  • Available as 10 mg solid and 10 mM in DMSO (1 mL)
  • Storage: solid at 4°C, protected from light; in solvent -80°C for up to 6 months, -20°C for 1 month

Catalog No. 50-002-96782 Supplier Medchemexpress LLC Supplier No. HYN000810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Orcinol glucoside is a natural phenolic glycoside isolated from Curculigo orchioides and supplied as a research-grade analytical standard for in vitro studies. It modulates mesenchymal stem cell differentiation via Wnt/β-catenin signaling and is intended for research use only.

  • High purity: 99.57%
  • Molecular weight 286.28 g·mol⁻¹
  • Chemical formula C13H18O7
  • Available as 10 mg solid and 10 mM in DMSO (1 mL)
  • Storage: solid at 4°C, protected from light; in solvent -80°C for up to 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.