Learn More
Selleck Chemical LLC Orcinol glucoside S3828-5mg
SDP
Catalog No. 503111451
Encompass_Preferred
Orcinol glucoside (OG Sakakin) is an active constituent isolated from the rhizomes of Curculigo orchioides Gaertn It shows potent antioxidative and anxiolytic activities without sedative effects
Catalog No. 50-311-1451 Supplier Selleck Chemical LLC Supplier No. S38285MG
Orcinol glucoside (OG Sakakin) is an active constituent isolated from the rhizomes of Curculigo orchioides Gaertn It shows potent antioxidative and anxiolytic activities without sedative effects