Learn More
Medchemexpress LLC Oregon green 488 azide | 2648246-11-9 | 98.0% | 637.67 | C33H37F2N5O6 | 10 MG
SDP
Catalog No. 5000296986
Encompass_Preferred
Oregon Green 488 azide is a green-fluorescent azide reagent used to label alkyne-modified biomolecules via copper-catalyzed azide-alkyne cycloaddition (click chemistry). It is supplied as a solid dye with high purity for biochemical assays and fluorescence labeling.
  • Bright green-fluorescent azide probe for click chemistry.
  • Reacts selectively with terminal alkynes via copper-catalyzed azide-alkyne cycloaddition.
  • High purity (98.0%) suitable for biochemical assays.
  • Solid, orange to red appearance for easy handling and storage.
  • Storage: sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in multiple pack sizes, including 10 MG.
Catalog No. 50-002-96986 Supplier Medchemexpress LLC Supplier No. HYD276910MG
Oregon Green 488 azide is a green-fluorescent azide reagent used to label alkyne-modified biomolecules via copper-catalyzed azide-alkyne cycloaddition (click chemistry). It is supplied as a solid dye with high purity for biochemical assays and fluorescence labeling.
  • Bright green-fluorescent azide probe for click chemistry.
  • Reacts selectively with terminal alkynes via copper-catalyzed azide-alkyne cycloaddition.
  • High purity (98.0%) suitable for biochemical assays.
  • Solid, orange to red appearance for easy handling and storage.
  • Storage: sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in multiple pack sizes, including 10 MG.