Learn More
Invitrogen™ Oregon Green™ 488-X, Succinimidyl Ester, 6-isomer
Description
The amine-reactive Oregon Green™ 488-X, succinimidyl ester can be used to can be used to create green fluorescent bioconjugates with excitation/emission maxima ∼496/524 nm. This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa (pKa ∼ 4.7 versus 6.4 for fluorescein), making its fluorescence essentially pH insensitive in the physiological pH range. This reactive dye contains an additional seven-atom aminohexanoyl spacer ('X') between the fluorophore and the succinimidyl ester group. This spacer helps to separate the fluorophore from its point of attachment, potentially reducing the interaction of the fluorophore with the biomolecule to which it is conjugated.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Succinimidyl Ester |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | Oregon Green™ 488 |
| Label Type | Classic Dyes |
| Product Line | Oregon Green |
| Emission | 524 |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.