Learn More
Cayman Chemical Oregon Green™ 488, Purity: ≥95%, CAS: 195136-58-4, 25 mg
SDP
Catalog No. 502909981
Encompass_Preferred
A fluorinated fluorescein dye; has minimal pH dependence at neutral pH, but due to its low pKa of 4.8, it can be used as an indicator for intracellular pH; less susceptible to photobleaching than fluorescein; ex/em = 490/514 nm, respectively
Catalog No. 50-290-9981 Supplier Cayman Chemical Supplier No. 3537225 MG
A fluorinated fluorescein dye; has minimal pH dependence at neutral pH, but due to its low pKa of 4.8, it can be used as an indicator for intracellular pH; less susceptible to photobleaching than fluorescein; ex/em = 490/514 nm, respectively