Learn More
Cayman Chemical Oregon Green488 50mg
SDP
Catalog No. 502913988
Encompass_Preferred
A fluorinated fluorescein dye; has minimal pH dependence at neutral pH, but due to its low pKa of 4.8, it can be used as an indicator for intracellular pH; less susceptible to photobleaching than fluorescein; ex/em = 490/514 nm, respectively
Catalog No. 50-291-3988 Supplier Cayman Chemical Supplier No. 3537250 MG
Add to Cart
Add to Cart
A fluorinated fluorescein dye; has minimal pH dependence at neutral pH, but due to its low pKa of 4.8, it can be used as an indicator for intracellular pH; less susceptible to photobleaching than fluorescein; ex/em = 490/514 nm, respectively