Learn More
MSC Oreq Corporation Cyanuric Acid, 55 lb, Bag
Catalog No. 502839904
Encompass
Cyanuric Acid is a high-quality water conditioner and stabilizer designed to control chlorine loss in pools and spas due to sun exposure. It comes in a fine powder or granular form, stabilizes chlorine by reducing UV degradation, and improves sanitizer performance and lifespan.
  • Stabilizes chlorine by reducing UV degradation
  • Improves sanitizer performance and lifespan
  • Available in child-safe, resealable pouches
  • Suitable for outdoor pools and spas
  • Reduces chlorine consumption
  • Supports balanced water chemistry
Catalog No. 50-283-9904 Supplier MSC Supplier No. 90607573
Cyanuric Acid is a high-quality water conditioner and stabilizer designed to control chlorine loss in pools and spas due to sun exposure. It comes in a fine powder or granular form, stabilizes chlorine by reducing UV degradation, and improves sanitizer performance and lifespan.
  • Stabilizes chlorine by reducing UV degradation
  • Improves sanitizer performance and lifespan
  • Available in child-safe, resealable pouches
  • Suitable for outdoor pools and spas
  • Reduces chlorine consumption
  • Supports balanced water chemistry