Learn More
Cayman Chemical Orexin 1 Receptor Reporter Assay Kit, 1 ea
SDP
Catalog No. 502895554
Encompass_Preferred
Cayman's OX1R Reporter Assay Kit consists of a 96-well plate coated with a transfection complex containing DNA constructs for OX1R, a recombinant G-protein, and a cAMP response element regulated Secreted Alkaline Phosphatase (SEAP) reporter (OX1R Reverse Transfection Strip Plate). Cells grown on the transfection complex will express OX1R at the cell surface and the recombinant G-protein inside the cell. Binding of agonists to OX1R initiates a signal transduction cascade resulting in expression of SEAP which is secreted into the cell culture medium. Aliquots of culture medium are removed at time intervals beginning at about 16 hours and SEAP activity is measured following addition of a luminescence-based alkaline phosphatase substrate provided in the kit. The kit is easy to use and can be easily adapted to high throughput screening for therapeutic compounds regulating activation of OX1R. A known OX1R agonist, Orexin A, is.
Catalog No. 50-289-5554 Supplier Cayman Chemical Supplier No. 6002401 EA
Cayman's OX1R Reporter Assay Kit consists of a 96-well plate coated with a transfection complex containing DNA constructs for OX1R, a recombinant G-protein, and a cAMP response element regulated Secreted Alkaline Phosphatase (SEAP) reporter (OX1R Reverse Transfection Strip Plate). Cells grown on the transfection complex will express OX1R at the cell surface and the recombinant G-protein inside the cell. Binding of agonists to OX1R initiates a signal transduction cascade resulting in expression of SEAP which is secreted into the cell culture medium. Aliquots of culture medium are removed at time intervals beginning at about 16 hours and SEAP activity is measured following addition of a luminescence-based alkaline phosphatase substrate provided in the kit. The kit is easy to use and can be easily adapted to high throughput screening for therapeutic compounds regulating activation of OX1R. A known OX1R agonist, Orexin A, is.