Learn More
Cayman Chemical Orexin 2 Receptor Reporter Assay Kit, 1 ea
SDP
Catalog No. 502895607
Encompass_Preferred
Cayman's OX2R Reporter Assay Kit consists of a 96-well plate coated with both OX2R and Secreted Alkaline Phosphatase (SEAP) reporter constructs (OX2R Reverse Transfection Strip Plate). Cells grown on the transfection complex will express OX2R at the cell surface and the recombinant G-protein inside the cell. Binding of agonists to OX2R initiates a signal transduction cascade resulting in expression of SEAP which is secreted into the cell culture medium. Aliquots of culture medium are removed at time intervals beginning at about 16 hours and SEAP activity is measured following addition of a luminescence-based alkaline phosphatases substrate provided in the kit. The kit is easy to use and can be easily adapted to high throughput screening for therapeutic compounds regulating activation of OX2R. A known OX2R agonist, Orexin A, is included in the kit for use as a positive control. The kit provides sufficient reagent to measure.
Catalog No. 50-289-5607 Supplier Cayman Chemical Supplier No. 6002501 EA
Cayman's OX2R Reporter Assay Kit consists of a 96-well plate coated with both OX2R and Secreted Alkaline Phosphatase (SEAP) reporter constructs (OX2R Reverse Transfection Strip Plate). Cells grown on the transfection complex will express OX2R at the cell surface and the recombinant G-protein inside the cell. Binding of agonists to OX2R initiates a signal transduction cascade resulting in expression of SEAP which is secreted into the cell culture medium. Aliquots of culture medium are removed at time intervals beginning at about 16 hours and SEAP activity is measured following addition of a luminescence-based alkaline phosphatases substrate provided in the kit. The kit is easy to use and can be easily adapted to high throughput screening for therapeutic compounds regulating activation of OX2R. A known OX2R agonist, Orexin A, is included in the kit for use as a positive control. The kit provides sufficient reagent to measure.