Learn More
Phoenix Pharmaceuticals Inc Orexin A (Human, Rat, Mouse, Porcine, Ovine, Bovine, Monkey), 100ug
Supplier:  Phoenix Pharmaceuticals Inc 00330
Encompass
Sequence: pGlu-Pro-Leu-Pro-Asp-Cys-Cys-Arg-Gln-Lys-Thr-Cys-Ser-Cys-Arg-Leu-Tyr-Glu-Leu-Leu-His-Gly-Ala-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Leu-NH2 Disulfide bridge(s): Cys6-Cys12 & Cys7-Cys14, Purity:≥ 95%, Molecular Weight: 3561.15, Soluble in water, Each vial contains 100 µg of NET peptide.
Catalog No. NC9427045