Learn More
Enzo Life Sciences Orexin A (human, bovine, rat, mouse) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX164001C100
Encompass_Preferred
Hypothalamic neuropeptide that stimulates food intake in rats with impressive potency. Binds specifically to the OX1 receptor. Formula: C152H243N47O44S4. MW: 3561.2. Purity: ≥98% (HPLC). Formulation: Lyophilized. Peptide content: 70-90%. Solubility: Soluble in DMSO; can be diluted with buffers or distilled water. Long Term Storage: -20°C.
Catalog No. 50-200-7849