Learn More
Cayman Chemical OrexIn A PosItIve Control 10uL
SDP
Supplier:  Cayman Chemical 60024210 UL
Encompass_Preferred
A peptide increases jejunum and ileum crypt depth and villus height without disrupting gut microbiota diversity in rats at 0.9 mg/kg four days per week decreases lapatinib-induced diarrhea severity and incidence in rats prevents lapatinib-induced loss of body weight in rats
Catalog No. 50-289-4361