Learn More
Cayman Chemical OrexIn A17-33humnmous 1mg
SDP
Catalog No. 502913870
Encompass_Preferred
A peptide orexin receptor 1 agonist; selectively induces calcium mobilization in CHO cells overexpressing human OX1R over CHO cells overexpressing human OX2R (EC50s = 8.29 and 187 nM, respectively)
Catalog No. 50-291-3870 Supplier Cayman Chemical Supplier No. 367651 MG
A peptide orexin receptor 1 agonist; selectively induces calcium mobilization in CHO cells overexpressing human OX1R over CHO cells overexpressing human OX2R (EC50s = 8.29 and 187 nM, respectively)