Learn More
Cayman Chemical OrexIn B amIdemouserat 1mg
SDP
Supplier:  Cayman Chemical 351281 MG
Encompass_Preferred
A hypothalamic neuropeptide; increases intracellular calcium mobilization in HEK293 cells expressing OX1R or OX2R (EC50s = 0.93 and 0.13 µM, respectively)
Catalog No. 50-291-2401