Learn More
Medchemexpress LLC HY-P1339A 1mg Medchemexpress, Orexin B, human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1339A1MG
Encompass_Preferred
Medchemexpress, HY-P1339A 1mg Orexin B, human (TFA) CAS: Orexin B, human (TFA) is an endogenous agonist at Orexin receptor with K i s of 420 and 36 nM for OX1 and OX2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4183