Learn More
ABclonal Technology Orexin B Rabbit mAb
SDP
Catalog No. 5000006664
Encompass_Preferred
This gene encodes a hypothalamic neuropeptide precursor protein that gives rise to two mature neuropeptides orexin A and orexin B by proteolytic processing Orexin A and orexin B which bind to orphan G-protein coupled receptors HCRTR1 and HCRTR2 function in the regulation of sleep and arousal This neuropeptide arrangement may also play a role in feeding behavior metabolism and homeostasis
Catalog No. 50-000-06664 Supplier ABclonal Technology Supplier No. A23383100UL
This gene encodes a hypothalamic neuropeptide precursor protein that gives rise to two mature neuropeptides orexin A and orexin B by proteolytic processing Orexin A and orexin B which bind to orphan G-protein coupled receptors HCRTR1 and HCRTR2 function in the regulation of sleep and arousal This neuropeptide arrangement may also play a role in feeding behavior metabolism and homeostasis