Learn More
Selleck Chemical LLC Orforglipron S9852-25MG
SDP
Catalog No. 5000024736
Encompass_Preferred
Orforglipron(LY3502970) is a non-peptidal agonist of glucagon-like peptide 1(GLP-1) receptor It can be used efficiently in glycemic control and weight reduction in type 2 diabetes mellitus (T2DM) obesity or both
Catalog No. 50-000-24736 Supplier Selleck Chemical LLC Supplier No. S985225MG
Orforglipron(LY3502970) is a non-peptidal agonist of glucagon-like peptide 1(GLP-1) receptor It can be used efficiently in glycemic control and weight reduction in type 2 diabetes mellitus (T2DM) obesity or both