Learn More
Enzo Life Sciences ORG-12962 (50mg). CAS: 210821-63-9
SDP
Supplier:  Enzo Life Sciences BMLNS7050050
Encompass_Preferred
Selective serotonin 5-HT2C agonist. Displays antiaversive effects in a panic-like anxiety rat model. May display appetite regulating properties similar to other 5-HT2C agonists. Alternative name: 1-[6-Chloro-5-trifluoromethyl)-2-pyridinyl]-piperazine. Purity: ≥95% (NMR). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1220