Learn More
Cayman Chemical ORG 12962hydrochlorIde 10mg
SDP
Catalog No. 502938791
Encompass_Preferred
A 5-HT2C receptor agonist (EC50 = 97.72 nM in a FLIPR-based intracellular calcium assay using CHO-K1 cells expressing the human receptors); selective for 5-HT2C over 5-HT2A and 5-HT2B receptors (EC50s = 419.87 and 524.81 nM, respectively)
Catalog No. 50-293-8791 Supplier Cayman Chemical Supplier No. 2767510 MG
A 5-HT2C receptor agonist (EC50 = 97.72 nM in a FLIPR-based intracellular calcium assay using CHO-K1 cells expressing the human receptors); selective for 5-HT2C over 5-HT2A and 5-HT2B receptors (EC50s = 419.87 and 524.81 nM, respectively)