Learn More
Cayman Chemical ORG 12962 (hydrochloride), Purity: ≥98%, CAS: 210821-63-9, 50 mg
SDP
Catalog No. 502940754
Encompass_Preferred
A 5-HT2C receptor agonist (EC50 = 97.72 nM in a FLIPR-based intracellular calcium assay using CHO-K1 cells expressing the human receptors); selective for 5-HT2C over 5-HT2A and 5-HT2B receptors (EC50s = 419.87 and 524.81 nM, respectively)
Catalog No. 50-294-0754 Supplier Cayman Chemical Supplier No. 2767550 MG
A 5-HT2C receptor agonist (EC50 = 97.72 nM in a FLIPR-based intracellular calcium assay using CHO-K1 cells expressing the human receptors); selective for 5-HT2C over 5-HT2A and 5-HT2B receptors (EC50s = 419.87 and 524.81 nM, respectively)