Learn More
Apexbio Technology LLC Org 27569 868273-06-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062308
Encompass_Preferred
Org 27569 (CAS 868273-06-7) is an allosteric modulator targeting the cannabinoid CB1 receptor It enhances the binding of the agonist CP55940 to purified CB1 receptors while simultaneously inhibiting receptor functionality In electrically-stimulated mouse vas deferens assays Org 27569 reduced WIN55212-induced contractions exhibiting a pEC50 of 8 24 0 12 and a maximal response (Emax) of approximately 45 4% Binding studies suggest its interaction at a putative allosteric site on CB1 receptors reflected by parameters such as a pKB of 5 67 0 23 and a Log of 1 14 0 17 Org 27569 serves as a tool compound for studying CB1 receptor allosteric modulation
Catalog No. 50-000-62308 Supplier Apexbio Technology LLC Supplier No. B14265.1
Org 27569 (CAS 868273-06-7) is an allosteric modulator targeting the cannabinoid CB1 receptor It enhances the binding of the agonist CP55940 to purified CB1 receptors while simultaneously inhibiting receptor functionality In electrically-stimulated mouse vas deferens assays Org 27569 reduced WIN55212-induced contractions exhibiting a pEC50 of 8 24 0 12 and a maximal response (Emax) of approximately 45 4% Binding studies suggest its interaction at a putative allosteric site on CB1 receptors reflected by parameters such as a pKB of 5 67 0 23 and a Log of 1 14 0 17 Org 27569 serves as a tool compound for studying CB1 receptor allosteric modulation