Learn More
Perkin Elmer US LLC Flared PVC MP2 Two-Stop Peristaltic Pump Tubing - 0.38 mm I.D., Orange/Green, Pkg. 12
Catalog No. NC0233590
Encompass_Preferred
MPP PVC Tubing Flared, 0.47mm, orange green, 12/pk. Flared tubing is easier to change and more reliably connected than non-flared small-bore tubing.
Catalog No. NC0233590 Supplier Perkin Elmer US LLC Supplier No. N8145197
MPP PVC Tubing Flared, 0.47mm, orange green, 12/pk. Flared tubing is easier to change and more reliably connected than non-flared small-bore tubing.