Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Org41841 | 301847-37-0 | 99.8% | 402.53 g/mol | C19H22N4O2S2 | 100 MG
SDP

Catalog No. 5000214470
Encompass_Preferred

Org41841 is a small-molecule partial agonist of the luteinizing hormone/choriogonadotropin receptor (LHCGR) and the thyroid-stimulating hormone receptor (TSHR), provided for research use in studies of GPCR signaling and cAMP-mediated receptor responses.

  • Partial agonist of LHCGR and TSHR, suitable for receptor pharmacology studies.
  • Molecular weight 402.53 g/mol.
  • Molecular formula C19H22N4O2S2.
  • CAS number 301847-37-0.
  • Purity 99.77% with supporting analytical data (COA, LCMS).
  • Available in multiple solid sizes and as a DMSO solution format for in vitro use.

Catalog No. 50-002-14470 Supplier Medchemexpress LLC Supplier No. HY100271100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Org41841 is a small-molecule partial agonist of the luteinizing hormone/choriogonadotropin receptor (LHCGR) and the thyroid-stimulating hormone receptor (TSHR), provided for research use in studies of GPCR signaling and cAMP-mediated receptor responses.

  • Partial agonist of LHCGR and TSHR, suitable for receptor pharmacology studies.
  • Molecular weight 402.53 g/mol.
  • Molecular formula C19H22N4O2S2.
  • CAS number 301847-37-0.
  • Purity 99.77% with supporting analytical data (COA, LCMS).
  • Available in multiple solid sizes and as a DMSO solution format for in vitro use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.