Learn More
Fine Science Tools Organ Holding Forceps, Stainless Steel, 14.5 cm
SDP
Catalog No. NC9590153
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 8 x 11mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm
Catalog No. NC9590153 Supplier Fine Science Tools Supplier No. 1103115
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 8 x 11mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm