Learn More
ORGANABIO LLC Human Peripheral Blood CD56 Natural Killer (NK) Cells / Cryopreserved / Single Donor / 25-32x vials of 1x10E7 cells/vial / OrganaBio LEUKOPAC - NK cells / Research Use Only
Catalog No. NC3997809
Encompass
Peripheral blood-derived CD56 NK cells from recallable / HLA-typed healthy donors isolated by negative selection. Includes immunophenotyping / infectious disease testing / and high-resolution HLA typing (Class I and II). Half leukopak processed into vials. Available in 1x10E7 cell vials. OrganaBio Human CD56 NK Cells are isolated from peripheral blood of normal / healthy adult donors by negative selection. Each lot is cryopreserved and characterized by immunophenotyping. Comprehensive donor characterization includes infectious disease testing panel and high-resolution HLA typing with full demographics.
Catalog No. NC3997809 Supplier ORGANABIO LLC Supplier No. NKPB010HB
Peripheral blood-derived CD56 NK cells from recallable / HLA-typed healthy donors isolated by negative selection. Includes immunophenotyping / infectious disease testing / and high-resolution HLA typing (Class I and II). Half leukopak processed into vials. Available in 1x10E7 cell vials. OrganaBio Human CD56 NK Cells are isolated from peripheral blood of normal / healthy adult donors by negative selection. Each lot is cryopreserved and characterized by immunophenotyping. Comprehensive donor characterization includes infectious disease testing panel and high-resolution HLA typing with full demographics.