Learn More
ORGANABIO LLC Human Peripheral Blood CD3 Pan T Cells / Cryopreserved / Single Donor / 30-38x vials of 2.5x10E7 cells/vial / OrganaBio LEUKOPAC - T cells / Research Use Only
Catalog No. NC3997804
Encompass
Peripheral blood-derived CD3 pan T cells from recallable / HLA-typed healthy donors isolated by negative selection. Includes immunophenotyping / infectious disease testing / and high-resolution HLA typing (Class I and II). Half leukopak processed into vials. Available in 2.5x10E7 cell vials. OrganaBio Human CD3 Pan T Cells are isolated from peripheral blood of normal / healthy adult donors by negative selection. Each lot is cryopreserved and characterized by immunophenotyping. Comprehensive donor characterization includes infectious disease testing panel and high-resolution HLA typing with full demographics.
Catalog No. NC3997804 Supplier ORGANABIO LLC Supplier No. TPB025HB
Peripheral blood-derived CD3 pan T cells from recallable / HLA-typed healthy donors isolated by negative selection. Includes immunophenotyping / infectious disease testing / and high-resolution HLA typing (Class I and II). Half leukopak processed into vials. Available in 2.5x10E7 cell vials. OrganaBio Human CD3 Pan T Cells are isolated from peripheral blood of normal / healthy adult donors by negative selection. Each lot is cryopreserved and characterized by immunophenotyping. Comprehensive donor characterization includes infectious disease testing panel and high-resolution HLA typing with full demographics.