Learn More
ORGANABIO LLC Human Cord Blood CD34 Hematopoietic Stem Cells (HSC) / Cryopreserved / Single Donor / 5x10E5 cells/vial / OrganaBio HEMATOPAC / Research Use Only
Catalog No. NC3997775
Encompass
Cryopreserved CD34 hematopoietic stem cells from fresh human cord blood. Includes infectious disease testing and immunophenotyping for CD34 HSC. Available in three vial sizes. OrganaBio Human Cord Blood CD34 Hematopoietic Stem Cells are isolated from fresh human cord blood units and cryopreserved to maintain high viability and functionality.Each lot is characterized by immunophenotyping and tested for a comprehensive infectious disease panel. Detailed donor demographics including HLA typing / age / sex / BMI / race / ethnicity / and blood type are provided.
Catalog No. NC3997775 Supplier ORGANABIO LLC Supplier No. HSCCB001
Cryopreserved CD34 hematopoietic stem cells from fresh human cord blood. Includes infectious disease testing and immunophenotyping for CD34 HSC. Available in three vial sizes. OrganaBio Human Cord Blood CD34 Hematopoietic Stem Cells are isolated from fresh human cord blood units and cryopreserved to maintain high viability and functionality.Each lot is characterized by immunophenotyping and tested for a comprehensive infectious disease panel. Detailed donor demographics including HLA typing / age / sex / BMI / race / ethnicity / and blood type are provided.