Learn More
ORGANABIO LLC Human Peripheral Blood Mononuclear Cells (PBMCs) / Cryopreserved / Single Donor / 5x10E7 cells/vial / OrganaBio LEUKOPAC - PBMC / Research Use Only
Catalog No. NC3997792
Encompass
Human peripheral blood mononuclear cells (PBMCs) from recallable / HLA-typed healthy adult donors isolated by density gradient centrifugation. Comprehensive characterization including immunophenotyping (CD45 / CD14 / CD19 / CD56 / CD3 / CD3CD4 / CD3CD8) / infectious disease testing / and high-resolution HLA typing (Class I and II). Multiple vial sizes available. OrganaBio Human PBMCs are isolated from peripheral blood of normal / healthy adult donors by density gradient centrifugation. Each lot is cryopreserved and characterized by immunophenotyping. Comprehensive donor characterization includes infectious disease testing panel and high-resolution HLA typing with full donor demographics (age / sex / BMI / race / ethnicity / blood type).
Catalog No. NC3997792 Supplier ORGANABIO LLC Supplier No. PBMCPB050
Human peripheral blood mononuclear cells (PBMCs) from recallable / HLA-typed healthy adult donors isolated by density gradient centrifugation. Comprehensive characterization including immunophenotyping (CD45 / CD14 / CD19 / CD56 / CD3 / CD3CD4 / CD3CD8) / infectious disease testing / and high-resolution HLA typing (Class I and II). Multiple vial sizes available. OrganaBio Human PBMCs are isolated from peripheral blood of normal / healthy adult donors by density gradient centrifugation. Each lot is cryopreserved and characterized by immunophenotyping. Comprehensive donor characterization includes infectious disease testing panel and high-resolution HLA typing with full donor demographics (age / sex / BMI / race / ethnicity / blood type).