Learn More
ORGANABIO LLC ORGANABIO LLC Peripheral Blood Mononuclear Cells, Human, Cryopreserved, 10 Million Cells
Supplier:  ORGANABIO LLC PBMCPB010
Encompass
These high-quality cryopreserved peripheral blood mononuclear cells (PBMCs) are suitable for diverse research applications, including immuno-oncology, vaccine development, and autoimmune disease studies. All collections are IRB-approved, ensuring ethical sourcing and reliability.
  • Cryopreserved peripheral blood mononuclear cells
  • 10 million cells per vial
  • Sourced from healthy adult peripheral blood (leukapheresis)
  • High post-thaw viability (≥90%)
  • Immunophenotyping (CD3+, CD4+, CD8+, CD45+, CD14+, CD19+, CD56+)
  • Comprehensive infectious disease panel tested
  • FDA Registered and IRB Approved
  • Certificate of Analysis provided
Catalog No. NC3997790