Learn More
ORGANABIO LLC Human Autoimmune Peripheral Blood Mononuclear Cells (PBMCs) / Cryopreserved / Single Donor / 5x10E7 cells/vial / OrganaBio LEUKOPAC - PBMC (Lupus / UC / Celiac / Crohn / RA) / Research Use Only
Supplier:  ORGANABIO LLC DPBMC050
Encompass
Human peripheral blood PBMCs from recallable autoimmune donors (SLE / Lupus / Crohn / UC / Celiac / RA / etc). Cryopreserved characterized with immunophenotyping (CD45 / CD14 / CD19 / CD56 / CD3 / CD3CD4 / CD3CD8) / infectious disease testing / and HLA typing. Available in 1x10E7 and 5x10E7 cell vials. OrganaBio Human disease state PBMCs are isolated from peripheral blood of autoimmune donors. Each lot is cryopreserved and characterized by immunophenotyping. Includes infectious disease testing and HLA typing with full donor demographics. Ideal for autoimmune disease research / target validation / and drug development studies.
Catalog No. NC3997796