Learn More
Enzo Life Sciences ORGANELLE-ID-RGB III assay kit (1Kit)
SDP
Catalog No. 502012597
Encompass_Preferred
Compared with other commercially available dyes for labeling Golgi bodies, the green dye component of the ORGANELLE-ID-RGB III detection reagent is more faithfully localized to the Golgi apparatus, with minimal staining of the endoplasmic reticulum. The red dye component of the reagent stains the endoplasmic reticulum with high fidelity and is specifically designed for use with green fluorescing probes. ORGANELLE-ID-RGB III assay kit is validated with human cervical carcinoma cell line HeLa, human T-lymphocyte cell line, Jurkat, canine kidney cell line MDCK, and human bone osteosarcoma epithelial cell line U2OS. The kit should also be suitable for identifying Golgi body and endoplasmic reticulum perturbing agents and thus can be a useful tool for examining the transport and recycling of molecules from the GA to ER in cellular secretory pathways. Applications: Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-2597 Supplier Enzo Life Sciences Supplier No. ENZ51032K100
Compared with other commercially available dyes for labeling Golgi bodies, the green dye component of the ORGANELLE-ID-RGB III detection reagent is more faithfully localized to the Golgi apparatus, with minimal staining of the endoplasmic reticulum. The red dye component of the reagent stains the endoplasmic reticulum with high fidelity and is specifically designed for use with green fluorescing probes. ORGANELLE-ID-RGB III assay kit is validated with human cervical carcinoma cell line HeLa, human T-lymphocyte cell line, Jurkat, canine kidney cell line MDCK, and human bone osteosarcoma epithelial cell line U2OS. The kit should also be suitable for identifying Golgi body and endoplasmic reticulum perturbing agents and thus can be a useful tool for examining the transport and recycling of molecules from the GA to ER in cellular secretory pathways. Applications: Fluorescence microscopy, Fluorescent detection .