Learn More
Cell Signaling Technology Organelle Localization IF Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. NC1565648
Encompass
The Organelle Localization IF Antibody Sampler Kit provides an economical means for identification of cellular organelles by fluorescence immunocytochemistry (IF-IC). This kit includes enough primary antibody to perform at least twenty IF-IC tests or two western blots with each antibody. Subcellular location knowledge may reveal potential roles in various cellular processes, and these antibodies may also be used as western blot controls for fractionated cell lysates.
  • Includes primary antibodies for PDI, PDCD8, RCAS1, EEA1, and LAMP1
  • Provides reagents for at least twenty IF-IC tests or two western blots per antibody
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol, and less than 0.02% sodium azide
  • Store at –20°C and do not aliquot
Catalog No. NC1565648 Supplier Cell Signaling Technology Supplier No. 8653T
The Organelle Localization IF Antibody Sampler Kit provides an economical means for identification of cellular organelles by fluorescence immunocytochemistry (IF-IC). This kit includes enough primary antibody to perform at least twenty IF-IC tests or two western blots with each antibody. Subcellular location knowledge may reveal potential roles in various cellular processes, and these antibodies may also be used as western blot controls for fractionated cell lysates.
  • Includes primary antibodies for PDI, PDCD8, RCAS1, EEA1, and LAMP1
  • Provides reagents for at least twenty IF-IC tests or two western blots per antibody
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol, and less than 0.02% sodium azide
  • Store at –20°C and do not aliquot