Learn More
ECM Bioscience Organelle Marker Immunocytochemistry Kit, ECM Biosciences
Catalog No. 502068772
Encompass_Preferred
The organelle market kit can be used to co-localize proteins to specific organelles in cells. The kit includes a panel of antibodies that detect the following cell organelles:Caveolae = Caveolin 1; Endoplasmic Reticulum = Calnexin; Endosomes = EEA1; Golgi = GM130; Mitochondria = Hsp60; Nucleus = Histone H2B. Applications: ICC, IHC; Species Reactivity: H, Rt, Ms
Catalog No. 50-206-8772 Supplier ECM Bioscience Supplier No. OK7670
The organelle market kit can be used to co-localize proteins to specific organelles in cells. The kit includes a panel of antibodies that detect the following cell organelles:Caveolae = Caveolin 1; Endoplasmic Reticulum = Calnexin; Endosomes = EEA1; Golgi = GM130; Mitochondria = Hsp60; Nucleus = Histone H2B. Applications: ICC, IHC; Species Reactivity: H, Rt, Ms