Learn More
Enzo Life Sciences ORGANELLE-ID-RGB reagent II (200µl)
SDP
Catalog No. 502012653
Encompass_Preferred
The ORGANELLE-ID-RGB reagent II is a mixture of cell-permeable red fluorescent lysosomal dye, green fluorescent endoplasmic reticulum dye and blue fluorescent nucleic acid dye. The staining pattern arising from the combination of these three dyes permits visualization of the target organelles by fluorescence/confocal microscopy. The reagent, supplied as a 500X solution, is sufficient for 1000 microscopy assays. The single-tube format makes this multi-organelle stain reagent easy to use. Purity: ≥93% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-2653 Supplier Enzo Life Sciences Supplier No. ENZ53008C200
The ORGANELLE-ID-RGB reagent II is a mixture of cell-permeable red fluorescent lysosomal dye, green fluorescent endoplasmic reticulum dye and blue fluorescent nucleic acid dye. The staining pattern arising from the combination of these three dyes permits visualization of the target organelles by fluorescence/confocal microscopy. The reagent, supplied as a 500X solution, is sufficient for 1000 microscopy assays. The single-tube format makes this multi-organelle stain reagent easy to use. Purity: ≥93% (HPLC). Long Term Storage: -80°C.