Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Organic Acids Kit Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022547 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2547 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2547 Supplier MilliporeSigma Supelco Supplier No. 47264
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Organic Acids Kit contains individually packaged acid standards in the quantities listed. The organic acids are prepared, tested, and individually packaged using rigorous manufacturing procedures.

TRUSTED_SUSTAINABILITY

Specifications

Flash Point 40°C
Physical Form Neat
UN Number UN 3316 9
Type Food and Cosmetic Component Standard
Quantity 1 Kit
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.