Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Organic Chemistry: Chemical Identification of Biomolecules

Shop All Aldon Products


Description
Includes
Materials for 15 groups: 25mL each: Iodine/Potassium Iodide solution, biuret reagent, Benedict's solution, 1.0% Sudan III stain and vegetable oil; 10g each: soluble starch, albumin and glucose; teacher's guide; and copy masters
- Learn to identify a positive test result for proteins using biuret reagent
- Examine the reaction between Benedict's solution and a simple sugar
- Use Iodine/Potassium Iodide to test for the presence of starch
- Test for the presence of lipids using a fat-soluble dye
Specifications
Specifications
| Certifications/Compliance | 49 CFR 173.4 |
| Color | White |
| Grade | 7 to 14 |
| Age | 13 to 20 |
| For Use With (Application) | To understand the importance of proteins, carbohydrates and lipids in living organisms |
| Includes | 25ml iodine/potassium iodide solution, biuret reagent, benedict's solution, 1% sudan iii stain and vegetable oil, 10g soluble starch, albumin and glucose, teacher guide and copy master |
| Quantity | 1 |
| Dimensions (L x W x H) | 9.75 x 6.25 x 4 in. |
| Class Size | 15 Lab Groups |
| Product Type | Chemical Identification of Biomolecule Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?