Learn More
Synthware™ Organic Chemistry Kit
GREENER_CHOICESDP
Catalog No. 31502140 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
31-502-140 24/40
31-502-137 14/20
31-502-139 19/22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-502-140 Supplier Synthware™ Supplier No. K162440
Add to Cart
Add to Cart
Preassembled Organic Chemistry Kit
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Quantity 1 Ea.
Standard Taper Joint 24/40
Product Type Organic Chemistry Kit