Learn More
Chemglass Life Sciences Organic Chemistry Labset Kit, 14/20
SDP
Catalog No. 50873333
Encompass_Preferred
Organic Chemistry Labset Kit, 14/20. Chemglass organic chemistry kits contain the basic glassware required to perform basic experiments such as vacuum or atmospheric distillation, reflux or reflux with addition, extraction as well as other organic experiments. Each kit contains the 14 glass items listed in a polypropylene box with die cut foam inserts. Box dimensions are 13-1/2" x 10-1/2" x 4-1/2".
Catalog No. 50-873-333 Supplier Chemglass Life Sciences Supplier No. CG980001
Organic Chemistry Labset Kit, 14/20. Chemglass organic chemistry kits contain the basic glassware required to perform basic experiments such as vacuum or atmospheric distillation, reflux or reflux with addition, extraction as well as other organic experiments. Each kit contains the 14 glass items listed in a polypropylene box with die cut foam inserts. Box dimensions are 13-1/2" x 10-1/2" x 4-1/2".