Learn More
Sigma Aldrich Fine Chemicals Biosciences Organic contaminants in ho
Catalog No. 502907303
Encompass
This Standard Reference Material (SRM) is intended for use in evaluating analytical methods for the determination of selected polycyclic aromatic hydrocarbons (PAHs) polychlorinated biphenyl (PCB) congeners chlorinated pesticides and polybrominated diphenyl ether (PBDE) congeners in house dust and similar matrices. All of the constituents for which certified reference and information values are provided in SRM 2585 were naturally present in the dust material before processing. A unit of SRM 2585 consists of one bottle containing approximately 10 g of house dust. For more information please refer to the COA and SDS. SRM 2585 SDS
Catalog No. 50-290-7303 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST2585
This Standard Reference Material (SRM) is intended for use in evaluating analytical methods for the determination of selected polycyclic aromatic hydrocarbons (PAHs) polychlorinated biphenyl (PCB) congeners chlorinated pesticides and polybrominated diphenyl ether (PBDE) congeners in house dust and similar matrices. All of the constituents for which certified reference and information values are provided in SRM 2585 were naturally present in the dust material before processing. A unit of SRM 2585 consists of one bottle containing approximately 10 g of house dust. For more information please refer to the COA and SDS. SRM 2585 SDS