Learn More
GFS Chemicals Inc ORGANIC HALOGEN REAGENT, (Sodium Biphenyl Solution) | 20 X 15 ML | GFS CHEMICALS, INC. Item #: 54101 | Bulk #: 5005
SDP
Catalog No. NC9748335
Encompass_Preferred
ORGANIC HALOGEN REAGENT, (Sodium Biphenyl Solution) | Chemical Formula : (C!6H!5)!2Na | CAS # : 5137-46-2 | Specifications : Concentration, approximately 1.2M, Halides (as Cl) (ppm) 8 ppm
Catalog No. NC9748335 Supplier GFS Chemicals Inc Supplier No. 54101
ORGANIC HALOGEN REAGENT, (Sodium Biphenyl Solution) | Chemical Formula : (C!6H!5)!2Na | CAS # : 5137-46-2 | Specifications : Concentration, approximately 1.2M, Halides (as Cl) (ppm) 8 ppm