Learn More
Thermo Scientific Peristaltic Pump Tubing | Orange/White | 1 EA
Supplier:  Thermo Scientific 842312052421
Encompass
Designed for durability, Compatible with all iCAP Series ICP-OES systems regardless of peristaltic pump size
Catalog No. NC1282592