Learn More
Absolute Standards Organic Nutrients 100ml
SDP
Catalog No. NC0891307
Encompass
Organic Nutrients comes in the amount of 100ml and it is composed of equal parts TKN (cas#56-40-6) and Total Phosphorus (cas#13408-09-08). The concentration is 1000ug/ml.
Catalog No. NC0891307 Supplier Absolute Standards Supplier No. 54152
Organic Nutrients comes in the amount of 100ml and it is composed of equal parts TKN (cas#56-40-6) and Total Phosphorus (cas#13408-09-08). The concentration is 1000ug/ml.