Learn More
Teknova Organic Sperm Wash Buffer. 1000mL, DNase Tested, Sterile. 8 Pack.
SDP
Catalog No. 502387871
Encompass
50mM Sodium Chloride, 10mM Tris-HCl, 10mM Ethylenediaminetetraacetic acid (EDTA), 2.0% SDS
Catalog No. 50-238-7871 Supplier Teknova Supplier No. O96158PK
Add to Cart
Add to Cart
50mM Sodium Chloride, 10mM Tris-HCl, 10mM Ethylenediaminetetraacetic acid (EDTA), 2.0% SDS