The organic vapor monitor provides a cost-effective and convenient method for personal air sampling of organic vapors. Designed for ease of use, it clips onto clothing without interfering with work activities and requires no batteries, hoses, or pumps. It utilizes diffusion to collect air samples with an accuracy of +/-25% at a 95% confidence level for many workplace organic vapors.
- State-of-the-art monitor design
- Simple, cost-effective, and convenient
- High sample capacity for sampling a broad range of organic vapors
- Easy to use; clips to shirt lapel, collar, or pocket
- Unobtrusive; small and lightweight
- Valid method for air sampling and analysis
- No batteries, hoses, or pumps needed
- Contains a single charcoal sorbent wafer
- Helps in determining appropriate respirator and cartridge