Learn More
Dentec Safety™ Organic Vapor Particulate Cartridge
SDP
Catalog No. 19222508 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Quantity:
100/Box
6/Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-222-508 100/Box
19-222-507 6/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-508 Supplier Dentec Safety™ Supplier No. 158T20B100
Add to Cart
Add to Cart
Dentec Safety™ Organic Vapor Particulate Cartridge features unique space plenums that allow users to access the entire filter material reducing breathing resistance, providing longer cartrdige life, reducing costs and improving productivity.
  • Offset connector allows the user to rotate the cartridge into 3 different positions to improve visibility and allow faceshields or welding helmets to be worn without interference
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Material Plastic
Product Type Respirator Accessories
For Use With (Equipment) Masks
Includes Cartridges and filters
Quantity 100/Box