Learn More
Perkin Elmer US LLC Organics High Sensitivity Plastic Nebulizer Tantalum Needle Assembly for PinAAcle 900
Catalog No. 502112810
Encompass_Preferred
The high sensitivity plastic nebulizer provides exceptional sensitivity, precision and corrosion resistance and can be used to aspirate almost all solutions, even those solutions containing a high percentage of acids.
Catalog No. 50-211-2810 Supplier Perkin Elmer US LLC Supplier No. N3160137
The high sensitivity plastic nebulizer provides exceptional sensitivity, precision and corrosion resistance and can be used to aspirate almost all solutions, even those solutions containing a high percentage of acids.