Learn More
Perkin Elmer US LLC Organics High Sensitivity Plastic Nebulizer Complete Assembly with Pt/Ir Needle for PinAAcle
Supplier:  Perkin Elmer US LLC N3160159
Encompass_Preferred
A complete High-Sensitivity plastic body nebulizer featuring Kalrez o-rings and Tantalum needle. The body is fabricated of inert plastic materials for maximum performance and corrosion resistance. This high-sensitivity nebulizer with Pt/Ir needle is for most solutions, but not for precious metal samples containing aqua regia. This assembly includes: Impact Bead, Qty. 1 (B0505086) Spacer, Qty. 1 (B3150531) Solvent Resistant O-Rings for Nebulizer Body, Qty. 2 (09200276) Plastic Nebulizer Body, Qty. 1 Spring, Qty. 2 (09920483) Locking Ring, Qty. 1 (N3151205) Solvent Resistant O-Ring for Capillary Needle Assembly, Qty. 1 (09200272) Solvent Resistant Pt/Ir Capillary Needle Assembly, Qty. 1 (N3160154) Fine Adjustment Nut, Qty. 1 (B3150462) Sample Tubing, Qty. 1 (B3150458) This nebulizer is compatible with the PerkinElmer PinAAcle 500/900F/900H/900T/900Z AA instruments.
Catalog No. 50-210-4361